The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128588.3 | NP_001122060.3 | 7 - 8 | 1057 | 80 |
NM_001146036.2 | NP_001139508.2 | 7 - 8 | 986 | 80 | |
NM_001146037.1 | NP_001139509.1 | 5 - 6 | 1154 | 80 | |
NM_001308278.1 | NP_001295207.1 | 5 - 6 | 731 | 80 | |
NM_001308279.1 | NP_001295208.1 | 4 - 5 | 541 | 80 | |
NM_015865.6 | NP_056949.4 | 6 - 7 | 899 | 80 | |
XM_005258329.1 | XP_005258386.1 | 7 - 8 | 1033 | 80 | |
XM_005258333.1 | XP_005258390.1 | 3 - 4 | 447 | 80 | |
XM_006722526.3 | XP_006722589.1 | 7 - 8 | 1034 | 80 | |
XM_011526142.1 | XP_011524444.1 | 6 - 7 | 1814 | 80 | |
XM_011526144.1 | XP_011524446.1 | 7 - 8 | 1033 | 80 | |
GenBank mRNA | AF328892.1 | - | 4 - 5 | 457 | 80 |
AK091064.1 | - | 5 - 6 | 1382 | 80 | |
AK123681.1 | - | 4 - 5 | 1958 | 80 | |
AK127452.1 | - | 5 - 6 | 643 | 80 | |
AK289608.1 | - | 7 - 8 | 1000 | 80 | |
AK294129.1 | - | 5 - 6 | 670 | 80 | |
AK295268.1 | - | 4 - 5 | 480 | 80 | |
AV724913.1 | - | 1 - 2 | 223 | 80 | |
BC050539.1 | - | 6 - 7 | 825 | 80 | |
GQ502682.1 | - | 4 - 5 | 688 | 80 | |
KC677694.1 | - | 4 - 5 | 667 | 80 | |
L36121.1 | - | 4 - 5 | 685 | 86 | |
U35735.1 | - | 6 - 7 | 835 | 80 | |
Y19039.1 | - | 4 - 5 | 688 | 80 |