The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014786.3 | NP_055601.2 | 4 - 5 | 3592 | 60 |
XM_017018623.1 | XP_016874112.1 | 4 - 5 | 1983 | 60 | |
XM_017018624.1 | XP_016874113.1 | 3 - 4 | 1062 | 60 | |
GenBank mRNA | AB002335.1 | - | 4 - 5 | 2366 | 60 |
AF378754.1 | - | 4 - 5 | 3586 | 60 | |
BC137228.1 | - | 4 - 5 | 3592 | 60 |