The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017896.2 | NP_060366.1 | 1 - 2 | 165 | 89 |
GenBank mRNA | AK000609.1 | - | 1 - 2 | 101 | 89 |
AK299509.1 | - | 1 - 2 | 104 | 89 | |
BC032120.2 | - | 1 - 2 | 135 | 89 | |
CD676269.1 | - | 1 - 2 | 197 | 89 |