The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014727.2 | NP_055542.1 | 5 - 6 | 2722 | 97 |
XM_011527561.2 | XP_011525863.2 | 5 - 6 | 2593 | 97 | |
XM_011527562.2 | XP_011525864.1 | 5 - 6 | 2923 | 97 | |
XM_017027544.1 | XP_016883033.1 | 5 - 6 | 2923 | 97 | |
XM_017027545.1 | XP_016883034.1 | 4 - 5 | 2636 | 97 | |
GenBank mRNA | AB002302.3 | - | 3 - 4 | 1822 | 97 |
AF104918.1 | - | 3 - 4 | 736 | 97 | |
AF186605.1 | - | 5 - 6 | 2394 | 97 | |
AJ007041.1 | - | 5 - 6 | 2722 | 97 |