Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACSS3
Entrez Gene ID
79611
Gene Name
acyl-CoA synthetase short-chain family member 3
Gene Aliases
-
Chromosome Location
Chr.12: 81078018 - 81257795 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024560.2 NP_078836.1 14 - 15 1910 94
XM_005269150.2 XP_005269207.1 14 - 15 1844 94
XM_011538740.2 XP_011537042.1 14 - 15 1559 94
GenBank mRNA AK025616.1 - 14 - 15 1861 94
AK074267.1 - 4 - 5 334 94
AK074938.1 - 9 - 10 1091 94
AX764480.1 - 14 - 15 2060 94
AX764482.1 - 14 - 15 1816 94
BC009317.2 - 14 - 15 1910 94
BC015769.1 - 14 - 15 1910 94

View Full Product Details