Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACSS3
Entrez Gene ID
79611
Gene Name
acyl-CoA synthetase short-chain family member 3
Gene Aliases
-
Chromosome Location
Chr.12: 81078018 - 81257795 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024560.2 NP_078836.1 5 - 6 1012 119
XM_005269150.2 XP_005269207.1 5 - 6 946 119
XM_005269151.4 XP_005269208.1 5 - 6 1024 119
GenBank mRNA AK025616.1 - 5 - 6 963 119
AX764480.1 - 5 - 6 1162 119
AX764482.1 - 5 - 6 918 119
BC009317.2 - 5 - 6 1012 119
BC015769.1 - 5 - 6 1012 119

View Full Product Details