The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145443.2 | NP_001138915.1 | 12 - 13 | 1337 | 98 |
NM_001282630.2 | NP_001269559.1 | 12 - 13 | 1450 | 98 | |
NM_001314063.1 | NP_001300992.1 | 12 - 13 | 1661 | 98 | |
NM_001323016.1 | NP_001309945.1 | 12 - 13 | 1337 | 98 | |
NM_001323017.1 | NP_001309946.1 | 12 - 13 | 1711 | 98 | |
NM_004566.3 | NP_004557.1 | 12 - 13 | 1661 | 98 | |
XM_005252464.1 | XP_005252521.1 | 12 - 13 | 1661 | 98 | |
XM_011519493.1 | XP_011517795.1 | 11 - 12 | 1556 | 98 | |
XM_017016325.1 | XP_016871814.1 | 12 - 13 | 1661 | 98 | |
XM_017016326.1 | XP_016871815.1 | 11 - 12 | 1604 | 98 | |
XM_017016327.1 | XP_016871816.1 | 12 - 13 | 1413 | 98 | |
XM_017016328.1 | XP_016871817.1 | 11 - 12 | 1556 | 98 | |
XM_017016329.1 | XP_016871818.1 | 11 - 12 | 1604 | 98 | |
GenBank mRNA | AF056320.1 | - | 13 - 14 | 1321 | 98 |
AF109735.1 | - | 12 - 13 | 1391 | 98 | |
AK291263.1 | - | 12 - 13 | 1606 | 98 | |
AK303039.1 | - | 12 - 13 | 1270 | 98 | |
AK304450.1 | - | 12 - 13 | 1382 | 98 | |
BC037258.1 | - | 12 - 13 | 1262 | 98 | |
BC040482.1 | - | 12 - 13 | 1606 | 98 | |
D49817.1 | - | 12 - 13 | 1295 | 98 | |
L77662.1 | - | 12 - 13 | 1615 | 98 |