Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PFKFB3
Entrez Gene ID
5209
Gene Name
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3
Gene Aliases
IPFK2, PFK2, iPFK-2
Chromosome Location
Chr.10: 6144802 - 6254648 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145443.2 NP_001138915.1 8 - 9 895 99
NM_001282630.2 NP_001269559.1 8 - 9 1008 99
NM_001314063.1 NP_001300992.1 8 - 9 1219 99
NM_001323016.1 NP_001309945.1 8 - 9 895 99
NM_001323017.1 NP_001309946.1 8 - 9 1269 99
NM_004566.3 NP_004557.1 8 - 9 1219 99
NR_136554.1 8 - 9 945 99
XM_005252464.1 XP_005252521.1 8 - 9 1219 99
XM_011519493.1 XP_011517795.1 8 - 9 1219 99
XM_017016325.1 XP_016871814.1 8 - 9 1219 99
XM_017016326.1 XP_016871815.1 7 - 8 1162 99
XM_017016327.1 XP_016871816.1 8 - 9 971 99
XM_017016328.1 XP_016871817.1 8 - 9 1219 99
XM_017016329.1 XP_016871818.1 7 - 8 1162 99
GenBank mRNA AF056320.1 - 9 - 10 879 99
AF109735.1 - 8 - 9 949 99
AK131307.1 - 8 - 9 905 99
AK291263.1 - 8 - 9 1164 99
AK304450.1 - 8 - 9 940 99
BC037258.1 - 8 - 9 820 99
BC040482.1 - 8 - 9 1164 99
D49817.1 - 8 - 9 853 99
L77662.1 - 8 - 9 1173 99

View Full Product Details