Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO1
Entrez Gene ID
55107
Gene Name
anoctamin 1
Gene Aliases
DOG1, ORAOV2, TAOS2, TMEM16A
Chromosome Location
Chr.11: 70078302 - 70189545 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018043.5 NP_060513.5 16 - 17 1889 67
NR_030691.1 14 - 15 1647 67
XM_006718602.2 XP_006718665.1 17 - 18 1671 67
XM_006718604.2 XP_006718667.1 17 - 18 1786 67
XM_006718605.2 XP_006718668.1 14 - 15 1642 67
XM_011545121.2 XP_011543423.1 17 - 18 1663 67
XM_011545123.2 XP_011543425.1 17 - 18 1660 67
XM_011545124.2 XP_011543426.1 16 - 17 1651 67
XM_011545125.2 XP_011543427.1 16 - 17 1585 67
XM_011545126.2 XP_011543428.1 15 - 16 1573 67
XM_011545127.2 XP_011543429.1 15 - 16 1519 67
XM_011545128.2 XP_011543430.1 14 - 15 1507 67
XM_011545129.2 XP_011543431.1 17 - 18 1578 67
XM_011545131.2 XP_011543433.1 14 - 15 1515 67
XM_017017956.1 XP_016873445.1 16 - 17 1594 67
XM_017017957.1 XP_016873446.1 15 - 16 1585 67
GenBank mRNA AB845669.1 - 16 - 17 1584 67
AK001123.1 - 6 - 7 427 67
AK097619.1 - 14 - 15 1515 67
AK293088.1 - 16 - 17 1597 67
AL833582.1 - 16 - 17 1633 67
AY728143.1 - 16 - 17 1889 67
BC027590.2 - 10 - 11 763 67
BC033036.2 - 14 - 15 1603 67

View Full Product Details