The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006536.5 | NP_006527.1 | 11 - 12 | 2148 | 136 |
GenBank mRNA | AB026833.1 | - | 11 - 12 | 2027 | 136 |
AF043977.1 | - | 11 - 12 | 2094 | 136 | |
AF127980.1 | - | 11 - 12 | 1986 | 136 | |
AK290348.1 | - | 11 - 12 | 2053 | 136 | |
AK307937.1 | - | 11 - 12 | 2051 | 137 | |
BC041096.1 | - | 11 - 12 | 2049 | 136 |