The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006536.5 | NP_006527.1 | 12 - 13 | 2319 | 78 |
GenBank mRNA | AB026833.1 | - | 12 - 13 | 2198 | 78 |
AF043977.1 | - | 12 - 13 | 2265 | 78 | |
AF127980.1 | - | 12 - 13 | 2157 | 78 | |
AK290348.1 | - | 12 - 13 | 2224 | 78 | |
AK307937.1 | - | 12 - 13 | 2223 | 78 | |
BC041096.1 | - | 12 - 13 | 2220 | 78 |