The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006536.5 | NP_006527.1 | 6 - 7 | 1139 | 117 |
XM_011542448.2 | XP_011540750.1 | 6 - 7 | 1081 | 117 | |
GenBank mRNA | AB026833.1 | - | 6 - 7 | 1018 | 117 |
AF043977.1 | - | 6 - 7 | 1085 | 117 | |
AF127980.1 | - | 6 - 7 | 977 | 117 | |
AK290348.1 | - | 6 - 7 | 1044 | 117 | |
AK307937.1 | - | 6 - 7 | 1042 | 117 | |
BC041096.1 | - | 6 - 7 | 1040 | 117 |