The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037984.2 | NP_001033073.1 | 12 - 13 | 1877 | 94 |
NM_138570.3 | NP_612637.1 | 12 - 13 | 1877 | 94 | |
XM_005257019.1 | XP_005257076.1 | 12 - 13 | 1971 | 94 | |
XM_011524288.1 | XP_011522590.1 | 12 - 13 | 1992 | 94 | |
XM_011524289.1 | XP_011522591.1 | 12 - 13 | 1992 | 94 | |
XM_011524290.1 | XP_011522592.1 | 10 - 11 | 1247 | 94 | |
GenBank mRNA | AK093037.1 | - | 6 - 7 | 836 | 94 |
BC014642.1 | - | 12 - 13 | 1804 | 94 |