The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037984.2 | NP_001033073.1 | 3 - 4 | 679 | 59 |
NM_138570.3 | NP_612637.1 | 3 - 4 | 679 | 59 | |
XM_005257019.1 | XP_005257076.1 | 3 - 4 | 773 | 59 | |
XM_011524288.1 | XP_011522590.1 | 3 - 4 | 794 | 59 | |
XM_011524289.1 | XP_011522591.1 | 3 - 4 | 794 | 59 | |
XM_011524290.1 | XP_011522592.1 | 1 - 2 | 49 | 59 | |
GenBank mRNA | BC014642.1 | - | 3 - 4 | 606 | 59 |
BQ642219.1 | - | 3 - 4 | 646 | 59 |