The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052854.3 | NP_443086.1 | 4 - 5 | 1070 | 67 |
XM_006718380.3 | XP_006718443.1 | 4 - 5 | 1047 | 67 | |
GenBank mRNA | AB063321.1 | - | 4 - 5 | 691 | 67 |
AK314072.1 | - | 4 - 5 | 1048 | 67 | |
BC014097.1 | - | 4 - 5 | 713 | 67 | |
BC015781.1 | - | 4 - 5 | 699 | 67 |