Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PICALM
Entrez Gene ID
8301
Gene Name
phosphatidylinositol binding clathrin assembly protein
Gene Aliases
CALM, CLTH, LAP
Chromosome Location
Chr.11: 85957171 - 86069881 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008660.2 NP_001008660.1 11 - 12 1475 91
NM_001206946.1 NP_001193875.1 11 - 12 1475 91
NM_001206947.1 NP_001193876.1 11 - 12 1190 91
NM_007166.3 NP_009097.2 11 - 12 1475 91
XM_005274322.2 XP_005274379.1 11 - 12 1459 91
XM_005274323.2 XP_005274380.1 11 - 12 1459 91
XM_005274324.2 XP_005274381.1 11 - 12 1459 91
XM_005274325.2 XP_005274382.1 11 - 12 1459 91
XM_005274326.2 XP_005274383.1 11 - 12 1459 91
XM_005274327.2 XP_005274384.1 11 - 12 1459 91
XM_005274328.2 XP_005274385.1 11 - 12 1459 91
XM_005274329.3 XP_005274386.1 11 - 12 1459 91
XM_005274330.2 XP_005274387.1 11 - 12 1459 91
XM_005274331.2 XP_005274388.1 11 - 12 1459 91
XM_005274332.2 XP_005274389.1 11 - 12 1459 91
XM_005274333.2 XP_005274390.1 11 - 12 1459 91
XM_005274334.2 XP_005274391.1 11 - 12 1459 91
XM_005274335.2 XP_005274392.1 11 - 12 1459 91
XM_005274336.2 XP_005274393.1 11 - 12 1459 91
XM_005274337.2 XP_005274394.1 11 - 12 1459 91
XM_005274340.2 XP_005274397.1 11 - 12 1459 91
XM_006718699.2 XP_006718762.1 11 - 12 1459 91
XM_006718700.2 XP_006718763.1 11 - 12 1459 91
XM_006718701.3 XP_006718764.1 11 - 12 1459 91
XM_011545293.1 XP_011543595.1 11 - 12 1459 91
XM_017018381.1 XP_016873870.1 11 - 12 1459 91
XM_017018382.1 XP_016873871.1 11 - 12 1459 91
XM_017018383.1 XP_016873872.1 11 - 12 1459 91
XM_017018384.1 XP_016873873.1 11 - 12 1459 91
XM_017018385.1 XP_016873874.1 11 - 12 1459 91
XM_017018386.1 XP_016873875.1 11 - 12 1459 91
XM_017018387.1 XP_016873876.1 11 - 12 1459 91
XM_017018388.1 XP_016873877.1 11 - 12 1459 91
GenBank mRNA AB210017.1 - 11 - 12 1475 91
AB451308.1 - 11 - 12 1158 91
AK291414.1 - 12 - 13 1342 91
AK297062.1 - 3 - 4 360 91
AK300275.1 - 11 - 12 1167 91
BC048259.1 - 11 - 12 1236 91
BC064357.1 - 11 - 12 1437 91
BC073961.1 - 11 - 12 1311 91
U45976.1 - 11 - 12 1305 91

View Full Product Details