The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008660.2 | NP_001008660.1 | 13 - 14 | 1679 | 101 |
NM_001206946.1 | NP_001193875.1 | 14 - 15 | 1808 | 101 | |
NM_001206947.1 | NP_001193876.1 | 13 - 14 | 1394 | 101 | |
NM_007166.3 | NP_009097.2 | 14 - 15 | 1829 | 101 | |
XM_005274322.2 | XP_005274379.1 | 14 - 15 | 1813 | 101 | |
XM_005274324.2 | XP_005274381.1 | 14 - 15 | 1813 | 101 | |
XM_005274326.2 | XP_005274383.1 | 14 - 15 | 1813 | 101 | |
XM_005274327.2 | XP_005274384.1 | 14 - 15 | 1813 | 101 | |
XM_005274330.2 | XP_005274387.1 | 13 - 14 | 1663 | 101 | |
XM_005274331.2 | XP_005274388.1 | 14 - 15 | 1813 | 101 | |
XM_005274333.2 | XP_005274390.1 | 13 - 14 | 1663 | 101 | |
XM_005274337.2 | XP_005274394.1 | 14 - 15 | 1813 | 101 | |
XM_005274340.2 | XP_005274397.1 | 13 - 14 | 1663 | 101 | |
XM_006718699.2 | XP_006718762.1 | 14 - 15 | 1792 | 101 | |
XM_006718701.3 | XP_006718764.1 | 14 - 15 | 1792 | 101 | |
XM_017018382.1 | XP_016873871.1 | 13 - 14 | 1663 | 101 | |
XM_017018383.1 | XP_016873872.1 | 13 - 14 | 1663 | 101 | |
XM_017018386.1 | XP_016873875.1 | 13 - 14 | 1663 | 101 | |
GenBank mRNA | AB210017.1 | - | 14 - 15 | 1808 | 101 |
AB451308.1 | - | 13 - 14 | 1362 | 101 | |
AK291414.1 | - | 15 - 16 | 1696 | 101 | |
AK300275.1 | - | 13 - 14 | 1371 | 101 | |
BC048259.1 | - | 14 - 15 | 1590 | 101 | |
BC064357.1 | - | 13 - 14 | 1641 | 101 | |
BC073961.1 | - | 13 - 14 | 1515 | 101 | |
U45976.1 | - | 14 - 15 | 1659 | 101 |