The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002336.2 | NP_002327.2 | 18 - 19 | 4110 | 66 |
XM_006719078.3 | XP_006719141.1 | 18 - 19 | 4254 | 66 | |
XM_011520671.2 | XP_011518973.1 | 17 - 18 | 3860 | 66 | |
XM_017019304.1 | XP_016874793.1 | 16 - 17 | 3928 | 66 | |
GenBank mRNA | AB209683.1 | - | 17 - 18 | 3562 | 66 |
AF074264.1 | - | 18 - 19 | 4045 | 66 | |
AK074543.1 | - | 3 - 4 | 478 | 66 | |
AK074902.1 | - | 8 - 9 | 1647 | 66 | |
BC117136.1 | - | 18 - 19 | 4002 | 66 | |
BC126405.1 | - | 18 - 19 | 4002 | 66 | |
BC143725.1 | - | 18 - 19 | 3867 | 66 |