The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022735.3 | NP_073572.2 | 6 - 7 | 1135 | 72 |
GenBank mRNA | AB043587.2 | - | 6 - 7 | 1143 | 72 |
AK025520.1 | - | 6 - 7 | 1136 | 72 | |
AK314468.1 | - | 6 - 7 | 1136 | 72 | |
AY150218.1 | - | 6 - 7 | 1114 | 72 | |
BC034563.2 | - | 4 - 5 | 584 | 72 | |
BC060792.1 | - | 6 - 7 | 1131 | 72 |