The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018159.1 | NP_001018169.1 | 4 - 5 | 388 | 67 |
NM_001018160.1 | NP_001018170.1 | 2 - 3 | 186 | 67 | |
NM_001286500.1 | NP_001273429.1 | 3 - 4 | 299 | 67 | |
NM_003905.3 | NP_003896.1 | 3 - 4 | 290 | 67 | |
GenBank mRNA | AK097680.1 | - | 3 - 4 | 299 | 67 |
AK298159.1 | - | 4 - 5 | 337 | 67 | |
AK307677.1 | - | 2 - 3 | 178 | 67 | |
AK312784.1 | - | 3 - 4 | 290 | 67 | |
AL136798.1 | - | 3 - 4 | 259 | 67 | |
AY197612.1 | - | 3 - 4 | 228 | 67 | |
BC000480.2 | - | 3 - 4 | 221 | 67 | |
BC013301.2 | - | 3 - 4 | 221 | 67 | |
BM045535.1 | - | 4 - 5 | 359 | 67 | |
BM785899.1 | - | 3 - 4 | 290 | 67 | |
U50939.1 | - | 3 - 4 | 286 | 67 |