The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018159.1 | NP_001018169.1 | 10 - 11 | 856 | 159 |
NM_001018160.1 | NP_001018170.1 | 8 - 9 | 654 | 159 | |
NM_001286500.1 | NP_001273429.1 | 9 - 10 | 767 | 159 | |
NM_003905.3 | NP_003896.1 | 9 - 10 | 758 | 159 | |
XM_005256215.1 | XP_005256272.1 | 8 - 9 | 728 | 159 | |
XM_011523422.2 | XP_011521724.1 | 7 - 8 | 570 | 159 | |
GenBank mRNA | AK097680.1 | - | 9 - 10 | 767 | 159 |
AK298159.1 | - | 10 - 11 | 805 | 159 | |
AK312784.1 | - | 9 - 10 | 758 | 159 | |
AL136798.1 | - | 9 - 10 | 727 | 159 | |
AY197612.1 | - | 9 - 10 | 696 | 159 | |
BC000480.2 | - | 9 - 10 | 689 | 159 | |
BC013301.2 | - | 9 - 10 | 689 | 159 | |
U50939.1 | - | 9 - 10 | 754 | 159 |