The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001194946.1 | NP_001181875.1 | 11 - 12 | 1815 | 89 |
NM_001194947.1 | NP_001181876.1 | 10 - 11 | 1758 | 89 | |
NM_001291157.1 | NP_001278086.1 | 10 - 11 | 1847 | 89 | |
NM_004953.4 | NP_004944.3 | 3 - 4 | 1303 | 89 | |
NM_182917.4 | NP_886553.3 | 9 - 10 | 1939 | 89 | |
NM_198241.2 | NP_937884.1 | 10 - 11 | 1794 | 89 | |
NM_198242.2 | NP_937885.1 | 6 - 7 | 1379 | 89 | |
NM_198244.2 | NP_937887.1 | 7 - 8 | 1556 | 89 | |
GenBank mRNA | AB210013.1 | - | 9 - 10 | 1933 | 89 |
AF104913.1 | - | 7 - 8 | 1415 | 89 | |
AF281070.1 | - | 9 - 10 | 1797 | 89 | |
AK096719.1 | - | 8 - 9 | 1444 | 89 | |
AK128378.1 | - | 9 - 10 | 1543 | 89 | |
AK226160.1 | - | 10 - 11 | 1728 | 89 | |
AK294567.1 | - | 8 - 9 | 1425 | 89 | |
AK299763.1 | - | 7 - 8 | 1484 | 89 | |
AK308319.1 | - | 5 - 6 | 1144 | 89 | |
AL832754.1 | - | 6 - 7 | 1268 | 89 | |
AY082886.1 | - | 9 - 10 | 1797 | 89 | |
BC140892.1 | - | 11 - 12 | 1657 | 89 | |
BC140896.1 | - | 10 - 11 | 1600 | 89 | |
BM475154.1 | - | - | 481 | 89 | |
BX647812.1 | - | 10 - 11 | 1707 | 89 | |
D12686.1 | - | 3 - 4 | 1303 | 89 |