Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHRD
Entrez Gene ID
8646
Gene Name
chordin
Gene Aliases
-
Chromosome Location
Chr.3: 184380073 - 184389835 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304472.1 NP_001291401.1 10 - 11 1456 58
NM_001304473.1 NP_001291402.1 11 - 12 1482 58
NM_001304474.1 NP_001291403.1 10 - 11 1427 58
NM_003741.3 NP_003732.2 10 - 11 1456 58
NR_130747.1 8 - 9 1614 58
XM_011513254.1 XP_011511556.1 10 - 11 1309 58
XM_011513256.1 XP_011511558.1 8 - 9 1465 58
XM_017007388.1 XP_016862877.1 9 - 10 1390 58
XM_017007389.1 XP_016862878.1 9 - 10 1390 58
XM_017007390.1 XP_016862879.1 9 - 10 1390 58
XM_017007391.1 XP_016862880.1 9 - 10 1390 58
XM_017007392.1 XP_016862881.1 10 - 11 1309 58
XM_017007393.1 XP_016862882.1 9 - 10 1390 58
XM_017007394.1 XP_016862883.1 8 - 9 1178 58
GenBank mRNA AF076612.1 - 8 - 9 868 58
AF209928.1 - 10 - 11 1456 58
AF209930.1 - 11 - 12 1482 58
AF283325.1 - 10 - 11 1427 58
AK074147.1 - 8 - 9 1367 58
AK293456.1 - 10 - 11 1178 58
AK296316.1 - 7 - 8 1213 58
AK300213.1 - 2 - 3 798 58
AY358926.1 - 10 - 11 1346 58
BC112345.1 - 10 - 11 1241 58

View Full Product Details