Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNK16
Entrez Gene ID
83795
Gene Name
potassium two pore domain channel subfamily K member 16
Gene Aliases
K2p16.1, TALK-1, TALK1
Chromosome Location
Chr.6: 39314566 - 39322992 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135105.1 NP_001128577.1 2 - 3 345 67
NM_001135106.1 NP_001128578.1 2 - 3 345 67
NM_001135107.1 NP_001128579.1 2 - 3 345 67
NM_032115.3 NP_115491.1 2 - 3 345 67
XM_011514935.2 XP_011513237.1 3 - 4 401 67
XM_017011346.1 XP_016866835.1 2 - 3 439 67
GenBank mRNA AY253145.1 - 2 - 3 331 67
AY253146.1 - 2 - 3 331 67
BC111860.1 - 2 - 3 331 67
EU978943.1 - 2 - 3 379 67

View Full Product Details