The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144715.3 | NP_653316.3 | 10 - 11 | 2129 | 148 |
XM_005264889.1 | XP_005264946.1 | 10 - 11 | 2129 | 148 | |
XM_011533382.1 | XP_011531684.1 | 9 - 10 | 2066 | 148 | |
XM_011533383.2 | XP_011531685.2 | 10 - 11 | 1237 | 148 | |
XM_011533385.2 | XP_011531687.1 | 11 - 12 | 1505 | 148 | |
XM_017005741.1 | XP_016861230.1 | 12 - 13 | 1689 | 148 | |
XM_017005743.1 | XP_016861232.1 | 11 - 12 | 1456 | 148 | |
XM_017005744.1 | XP_016861233.1 | 10 - 11 | 1368 | 148 | |
XM_017005745.1 | XP_016861234.1 | 11 - 12 | 1431 | 148 | |
GenBank mRNA | AK057929.1 | - | 1 - 2 | 143 | 148 |
AK122616.1 | - | 12 - 13 | 1665 | 148 | |
BC028198.1 | - | 10 - 11 | 2095 | 148 |