The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001323.1 | NP_001001323.1 | 14 - 15 | 2617 | 128 |
NM_001682.2 | NP_001673.2 | 14 - 15 | 2617 | 128 | |
XM_011538407.2 | XP_011536709.1 | 15 - 16 | 2792 | 128 | |
XM_017019357.1 | XP_016874846.1 | 14 - 15 | 2569 | 128 | |
XM_017019358.1 | XP_016874847.1 | 14 - 15 | 2569 | 128 | |
XM_017019359.1 | XP_016874848.1 | 14 - 15 | 2569 | 128 | |
GenBank mRNA | AB208896.1 | - | 10 - 11 | 1534 | 128 |
AK024895.1 | - | 8 - 9 | 1397 | 128 | |
J04027.1 | - | 14 - 15 | 2617 | 128 | |
M95541.1 | - | 14 - 15 | 2617 | 128 | |
M95542.1 | - | 10 - 11 | 1795 | 128 |