The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000458.3 | NP_000449.1 | 5 - 6 | 1404 | 89 |
NM_001165923.3 | NP_001159395.1 | 5 - 6 | 1326 | 89 | |
NM_001304286.1 | NP_001291215.1 | 5 - 6 | 1326 | 89 | |
XM_011525160.1 | XP_011523462.1 | 5 - 6 | 1385 | 89 | |
XM_011525161.1 | XP_011523463.1 | 5 - 6 | 1385 | 89 | |
XM_011525164.1 | XP_011523466.1 | 5 - 6 | 1307 | 89 | |
GenBank mRNA | AK290131.1 | - | 5 - 6 | 1385 | 89 |
AK296633.1 | - | 5 - 6 | 1307 | 89 | |
BC017714.1 | - | 5 - 6 | 1398 | 89 | |
BT007126.1 | - | 5 - 6 | 1210 | 89 | |
CR536572.1 | - | 5 - 6 | 1210 | 89 | |
HM116553.1 | - | 5 - 6 | 1266 | 89 | |
HM116554.1 | - | 5 - 6 | 1247 | 89 | |
HM116556.1 | - | 5 - 6 | 1266 | 89 | |
X58840.1 | - | 5 - 6 | 1404 | 89 |