The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164380.1 | NP_001157852.1 | 13 - 14 | 1869 | 163 |
NM_001164381.1 | NP_001157853.1 | 12 - 13 | 1738 | 163 | |
NM_001164383.1 | NP_001157855.1 | 8 - 9 | 1263 | 163 | |
GenBank mRNA | AF459097.1 | - | 11 - 12 | 1588 | 163 |
AF459098.1 | - | 10 - 11 | 1457 | 163 | |
AK002152.1 | - | 13 - 14 | 1782 | 163 | |
AK293496.1 | - | 8 - 9 | 1176 | 163 | |
BC008369.1 | - | 12 - 13 | 1651 | 163 |