The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015192.3 | NP_056007.1 | 22 - 23 | 2801 | 76 |
NM_182734.2 | NP_877398.1 | 22 - 23 | 2801 | 76 | |
XM_011529199.2 | XP_011527501.1 | 22 - 23 | 2761 | 76 | |
XM_011529200.2 | XP_011527502.1 | 20 - 21 | 2425 | 76 | |
XM_011529201.2 | XP_011527503.1 | 20 - 21 | 2235 | 76 | |
XM_011529203.2 | XP_011527505.1 | 6 - 7 | 737 | 76 | |
XM_017027752.1 | XP_016883241.1 | 22 - 23 | 2761 | 76 | |
XM_017027753.1 | XP_016883242.1 | 22 - 23 | 2388 | 76 | |
GenBank mRNA | AB011153.3 | - | 22 - 23 | 2801 | 76 |
AJ278313.1 | - | 22 - 23 | 2417 | 76 | |
AJ278314.1 | - | 22 - 23 | 2417 | 76 | |
AK023689.1 | - | 10 - 11 | 1566 | 76 | |
AK127693.1 | - | 20 - 21 | 2216 | 76 | |
AK299195.1 | - | 22 - 23 | 2436 | 76 | |
AL137267.1 | - | 14 - 15 | 1636 | 76 | |
AY004175.1 | - | 22 - 23 | 2399 | 76 | |
BC069420.1 | - | 22 - 23 | 2417 | 76 | |
BC117231.1 | - | 22 - 23 | 2472 | 76 |