The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015192.3 | NP_056007.1 | 23 - 24 | 2912 | 71 |
NM_182734.2 | NP_877398.1 | 23 - 24 | 2912 | 71 | |
XM_011529199.2 | XP_011527501.1 | 23 - 24 | 2872 | 71 | |
XM_011529200.2 | XP_011527502.1 | 21 - 22 | 2536 | 71 | |
XM_011529201.2 | XP_011527503.1 | 21 - 22 | 2346 | 71 | |
XM_011529203.2 | XP_011527505.1 | 7 - 8 | 848 | 71 | |
XM_017027752.1 | XP_016883241.1 | 23 - 24 | 2872 | 71 | |
XM_017027753.1 | XP_016883242.1 | 23 - 24 | 2499 | 71 | |
GenBank mRNA | AB011153.3 | - | 23 - 24 | 2912 | 71 |
AJ278313.1 | - | 23 - 24 | 2528 | 71 | |
AJ278314.1 | - | 23 - 24 | 2528 | 71 | |
AK023689.1 | - | 11 - 12 | 1677 | 71 | |
AK127693.1 | - | 21 - 22 | 2327 | 71 | |
AK299195.1 | - | 23 - 24 | 2547 | 71 | |
AL137267.1 | - | 15 - 16 | 1747 | 71 | |
AY004175.1 | - | 23 - 24 | 2510 | 71 | |
BC069420.1 | - | 23 - 24 | 2528 | 71 | |
BC117231.1 | - | 23 - 24 | 2583 | 71 |