The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015192.3 | NP_056007.1 | 9 - 10 | 1252 | 149 |
NM_182734.2 | NP_877398.1 | 9 - 10 | 1252 | 149 | |
XM_011529199.2 | XP_011527501.1 | 9 - 10 | 1212 | 149 | |
XM_011529200.2 | XP_011527502.1 | 7 - 8 | 876 | 149 | |
XM_011529201.2 | XP_011527503.1 | 7 - 8 | 686 | 149 | |
XM_017027752.1 | XP_016883241.1 | 9 - 10 | 1212 | 149 | |
XM_017027753.1 | XP_016883242.1 | 9 - 10 | 839 | 149 | |
GenBank mRNA | AB011153.3 | - | 9 - 10 | 1252 | 149 |
AJ278313.1 | - | 9 - 10 | 868 | 149 | |
AJ278314.1 | - | 9 - 10 | 868 | 149 | |
AK127693.1 | - | 7 - 8 | 667 | 149 | |
AK299195.1 | - | 9 - 10 | 887 | 149 | |
AY004175.1 | - | 9 - 10 | 850 | 149 | |
BC069420.1 | - | 9 - 10 | 868 | 149 | |
BC117231.1 | - | 9 - 10 | 923 | 149 |