The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000834.3 | NP_000825.2 | 11 - 12 | 2540 | 59 |
XM_005253351.3 | XP_005253408.1 | 2 - 3 | 267 | 59 | |
XM_011520628.2 | XP_011518930.1 | 12 - 13 | 3333 | 59 | |
XM_011520629.2 | XP_011518931.1 | 13 - 14 | 3140 | 59 | |
XM_017019219.1 | XP_016874708.1 | 12 - 13 | 2888 | 59 | |
GenBank mRNA | AB208850.1 | - | 8 - 9 | 1319 | 59 |
BC113618.1 | - | 11 - 12 | 2439 | 59 | |
BC113620.1 | - | 11 - 12 | 2439 | 59 | |
U11287.1 | - | 11 - 12 | 2571 | 59 | |
U88963.1 | - | 11 - 12 | 2540 | 59 | |
U90278.1 | - | 11 - 12 | 2570 | 59 |