The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130028.1 | NP_001123500.1 | 1 - 2 | 905 | 115 |
XM_005254151.3 | XP_005254208.1 | 1 - 2 | 905 | 115 | |
XM_011521205.2 | XP_011519507.1 | 1 - 2 | 905 | 115 | |
XM_011521206.2 | XP_011519508.1 | 1 - 2 | 905 | 115 | |
GenBank mRNA | AK091692.1 | - | 1 - 2 | 905 | 115 |
BI459843.1 | - | 1 - 2 | 402 | 115 |