The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281460.1 | NP_001268389.1 | 1 - 2 | 89 | 56 |
XM_017027860.1 | XP_016883349.1 | 1 - 2 | 92 | 56 | |
GenBank mRNA | DB023133.1 | - | 1 - 2 | 32 | 56 |