Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35C2
Entrez Gene ID
51006
Gene Name
solute carrier family 35 member C2
Gene Aliases
BA394O2.1, C20orf5, CGI-15, OVCOV1
Chromosome Location
Chr.20: 46349528 - 46364461 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281457.1 NP_001268386.1 7 - 8 932 77
NM_001281458.1 NP_001268387.1 8 - 9 871 77
NM_001281459.1 NP_001268388.1 7 - 8 869 77
NM_001281460.1 NP_001268389.1 8 - 9 1335 77
NM_015945.11 NP_057029.8 7 - 8 1003 77
NM_173073.3 NP_775096.1 7 - 8 940 77
NM_173179.3 NP_775271.1 7 - 8 1198 77
XM_011528831.1 XP_011527133.1 7 - 8 1181 77
XM_011528832.1 XP_011527134.1 7 - 8 1475 77
XM_011528833.1 XP_011527135.1 7 - 8 999 77
XM_011528836.1 XP_011527138.1 7 - 8 1084 77
XM_011528837.1 XP_011527139.1 7 - 8 918 77
XM_011528838.1 XP_011527140.1 5 - 6 1037 77
XM_017027856.1 XP_016883345.1 7 - 8 1898 77
XM_017027857.1 XP_016883346.1 7 - 8 1426 77
XM_017027858.1 XP_016883347.1 7 - 8 1336 77
XM_017027859.1 XP_016883348.1 7 - 8 1703 77
XM_017027860.1 XP_016883349.1 8 - 9 1143 77
XM_017027861.1 XP_016883350.1 7 - 8 1102 77
XM_017027862.1 XP_016883351.1 7 - 8 1041 77
GenBank mRNA AF132949.1 - 7 - 8 940 77
AF455052.1 - 7 - 8 958 77
AK222928.1 - 7 - 8 943 77
AK300207.1 - 7 - 8 886 77
AK300798.1 - 7 - 8 801 77
AK301022.1 - 8 - 9 813 77
AK315726.1 - 7 - 8 1165 77
BC014191.2 - 7 - 8 884 77
BC021138.1 - 7 - 8 1141 77
BC025277.1 - 7 - 8 1199 77

View Full Product Details