Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNASEH2B
Entrez Gene ID
79621
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
Chromosome Location
Chr.13: 50909678 - 50970462 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142279.2 NP_001135751.1 9 - 10 1144 61
GenBank mRNA AK124228.1 - 4 - 5 2810 61
BX328027.1 - 6 - 7 471 61
BX330672.1 - 1 - 2 61
CA309004.1 - 1 - 2 61

View Full Product Details