Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C14orf39
Entrez Gene ID
317761
Gene Name
chromosome 14 open reading frame 39
Gene Aliases
Six6os1
Chromosome Location
Chr.14: 60435954 - 60499773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174978.2 NP_777638.2 17 - 18 1667 189
XM_011536690.2 XP_011534992.1 18 - 19 1797 189
XM_017021247.1 XP_016876736.1 18 - 19 1795 189
XM_017021248.1 XP_016876737.1 16 - 17 1630 189
XM_017021249.1 XP_016876738.1 14 - 15 1362 189
XM_017021250.1 XP_016876739.1 13 - 14 1712 189
XM_017021251.1 XP_016876740.1 13 - 14 1313 189
GenBank mRNA AK098187.1 - 17 - 18 1652 189
AL832219.1 - 17 - 18 1717 189
BC125070.1 - 17 - 18 1638 189

View Full Product Details