Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C14orf39
Entrez Gene ID
317761
Gene Name
chromosome 14 open reading frame 39
Gene Aliases
Six6os1
Chromosome Location
Chr.14: 60435954 - 60499773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174978.2 NP_777638.2 3 - 4 214 163
GenBank mRNA AK098187.1 - 3 - 4 199 163
AL832219.1 - 3 - 4 264 163
BC125070.1 - 3 - 4 185 163
DB447488.2 - 3 - 4 233 163

View Full Product Details