The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256545.1 | NP_001243474.1 | 14 - 15 | 2156 | 60 |
NM_032446.2 | NP_115822.1 | 15 - 16 | 2207 | 60 | |
XM_011543694.1 | XP_011541996.1 | 14 - 15 | 1919 | 60 | |
XM_017009987.1 | XP_016865476.1 | 16 - 17 | 2452 | 60 | |
XM_017009988.1 | XP_016865477.1 | 7 - 8 | 1225 | 60 | |
GenBank mRNA | AB058676.1 | - | 15 - 16 | 2048 | 60 |
AK123568.1 | - | 16 - 17 | 2322 | 60 | |
BC152478.1 | - | 15 - 16 | 2048 | 60 | |
CR749437.1 | - | 14 - 15 | 2090 | 60 | |
DA738723.1 | - | 3 - 4 | 431 | 60 |