The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040876.1 | NP_001035809.1 | 2 - 3 | 546 | 114 |
NM_002940.2 | NP_002931.2 | 2 - 3 | 549 | 114 | |
GenBank mRNA | AB062293.1 | - | 2 - 3 | 223 | 114 |
AK315198.1 | - | 2 - 3 | 242 | 114 | |
BC016283.1 | - | 2 - 3 | 211 | 114 | |
BT009779.1 | - | 1 - 2 | 106 | 114 | |
X76388.1 | - | 2 - 3 | 223 | 114 |