The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040876.1 | NP_001035809.1 | 3 - 4 | 633 | 157 |
NM_002940.2 | NP_002931.2 | 3 - 4 | 636 | 157 | |
GenBank mRNA | AB062293.1 | - | 3 - 4 | 310 | 157 |
AK315198.1 | - | 3 - 4 | 329 | 157 | |
BC016283.1 | - | 3 - 4 | 298 | 157 | |
BT009779.1 | - | 2 - 3 | 193 | 157 | |
X76388.1 | - | 3 - 4 | 310 | 157 |