The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320610.1 | NP_001307539.1 | 5 - 6 | 594 | 66 |
NM_032343.2 | NP_115719.1 | 5 - 6 | 591 | 66 | |
XM_011513234.1 | XP_011511536.1 | 5 - 6 | 531 | 66 | |
XM_017007320.1 | XP_016862809.1 | 5 - 6 | 573 | 66 | |
XM_017007321.1 | XP_016862810.1 | 5 - 6 | 573 | 66 | |
GenBank mRNA | AK310910.1 | - | 5 - 6 | 573 | 66 |
AK311056.1 | - | 6 - 7 | 671 | 66 | |
BC006123.1 | - | 5 - 6 | 536 | 66 | |
DN992600.1 | - | 5 - 6 | 598 | 66 | |
JF264889.1 | - | 5 - 6 | 536 | 66 |