The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303542.2 | NP_001290471.2 | 13 - 13 | 8264 | 90 |
NM_032932.5 | NP_116321.2 | 15 - 15 | 8557 | 90 | |
GenBank mRNA | AJ314646.2 | - | 15 - 15 | 8247 | 90 |
AL359563.2 | - | 1 - 1 | 2860 | 90 | |
BC006240.2 | - | 1 - 1 | 690 | 90 | |
BC047674.2 | - | 1 - 1 | 1109 | 90 | |
BC094803.1 | - | 1 - 1 | 752 | 90 |