Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A27
Entrez Gene ID
9481
Gene Name
solute carrier family 25 member 27
Gene Aliases
UCP4
Chromosome Location
Chr.6: 46652915 - 46678190 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204051.1 NP_001190980.1 1 - 2 386 99
NM_001204052.1 NP_001190981.1 1 - 2 386 99
NM_004277.4 NP_004268.3 1 - 2 386 99
XM_005249483.2 XP_005249540.1 1 - 2 356 99
GenBank mRNA AF110532.1 - 1 - 2 108 99
AK291427.1 - 1 - 2 316 99
AK295233.1 - 1 - 2 369 99
AK302906.1 - 1 - 2 326 99
AK315940.1 - 1 - 2 297 99
AY358711.1 - 1 - 2 134 99
BC033091.1 - 1 - 2 359 99
BP310559.1 - 2 - 3 273 99
DA144842.1 - 1 - 2 360 99
DC314392.1 - 1 - 2 386 99
DR002070.1 - 1 - 2 308 99

View Full Product Details