The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243597.1 | NP_001230526.1 | 18 - 19 | 3481 | 67 |
NM_016952.4 | NP_058648.4 | 18 - 19 | 3481 | 67 | |
XM_011542862.2 | XP_011541164.1 | 18 - 19 | 3642 | 67 | |
XM_011542863.2 | XP_011541165.1 | 18 - 19 | 3646 | 67 | |
XM_011542864.2 | XP_011541166.1 | 18 - 19 | 3559 | 67 | |
XM_011542865.2 | XP_011541167.1 | 18 - 19 | 3442 | 67 | |
XM_011542866.2 | XP_011541168.1 | 18 - 19 | 3642 | 67 | |
XM_017017873.1 | XP_016873362.1 | 18 - 19 | 3478 | 67 | |
GenBank mRNA | AB209396.1 | - | 9 - 10 | 1870 | 67 |
AF004841.2 | - | 18 - 19 | 3509 | 67 | |
BC098583.1 | - | 18 - 19 | 3481 | 67 | |
BC114435.1 | - | 17 - 18 | 3385 | 67 |