The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243597.1 | NP_001230526.1 | 9 - 10 | 1980 | 69 |
NM_016952.4 | NP_058648.4 | 9 - 10 | 1980 | 69 | |
XM_011542862.2 | XP_011541164.1 | 9 - 10 | 2141 | 69 | |
XM_011542863.2 | XP_011541165.1 | 9 - 10 | 2145 | 69 | |
XM_011542864.2 | XP_011541166.1 | 9 - 10 | 2058 | 69 | |
XM_011542865.2 | XP_011541167.1 | 9 - 10 | 1941 | 69 | |
XM_011542866.2 | XP_011541168.1 | 9 - 10 | 2141 | 69 | |
XM_017017873.1 | XP_016873362.1 | 9 - 10 | 1977 | 69 | |
GenBank mRNA | AF004841.2 | - | 9 - 10 | 2008 | 69 |
BC098583.1 | - | 9 - 10 | 1980 | 69 | |
BC114435.1 | - | 8 - 9 | 1884 | 69 |