The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018060.2 | NP_001018070.1 | 15 - 16 | 1575 | 61 |
NM_001146288.1 | NP_001139760.1 | 15 - 16 | 1660 | 61 | |
NM_144704.2 | NP_653305.1 | 15 - 16 | 1575 | 61 | |
NR_027464.1 | 14 - 15 | 1532 | 61 | ||
GenBank mRNA | AK055035.1 | - | 15 - 16 | 1559 | 61 |
AK094844.1 | - | 15 - 16 | 1564 | 61 | |
AK126917.1 | - | 1 - 2 | 902 | 61 | |
AK295581.1 | - | 14 - 15 | 1435 | 61 | |
AK316042.1 | - | 14 - 15 | 1428 | 61 | |
AY927594.1 | - | 4 - 5 | 61 | ||
BC032485.1 | - | 15 - 16 | 1542 | 61 | |
CR456342.1 | - | 14 - 15 | 1394 | 61 |