The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018060.2 | NP_001018070.1 | 16 - 17 | 1699 | 70 |
NM_001146288.1 | NP_001139760.1 | 16 - 17 | 1784 | 70 | |
NM_144704.2 | NP_653305.1 | 16 - 17 | 1699 | 70 | |
NR_027464.1 | 15 - 16 | 1656 | 70 | ||
GenBank mRNA | AK055035.1 | - | 16 - 17 | 1683 | 70 |
AK094844.1 | - | 16 - 17 | 1688 | 70 | |
AK126917.1 | - | 2 - 3 | 1026 | 70 | |
AK295581.1 | - | 15 - 16 | 1559 | 70 | |
AK316042.1 | - | 15 - 16 | 1552 | 70 | |
AY927594.1 | - | 3 - 4 | 70 | ||
BC032485.1 | - | 16 - 17 | 1666 | 70 | |
CR456342.1 | - | 15 - 16 | 1518 | 70 |