The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018060.2 | NP_001018070.1 | 17 - 18 | 1793 | 65 |
NM_001146288.1 | NP_001139760.1 | 17 - 18 | 1878 | 65 | |
NM_144704.2 | NP_653305.1 | 17 - 18 | 1793 | 65 | |
NR_027464.1 | 16 - 17 | 1750 | 65 | ||
GenBank mRNA | AK055035.1 | - | 17 - 18 | 1777 | 65 |
AK094844.1 | - | 17 - 18 | 1782 | 65 | |
AK126917.1 | - | 3 - 4 | 1120 | 65 | |
AK295581.1 | - | 16 - 17 | 1653 | 65 | |
AK316042.1 | - | 16 - 17 | 1646 | 65 | |
AY927594.1 | - | 2 - 3 | 65 | ||
BC032485.1 | - | 17 - 18 | 1760 | 65 | |
CR456342.1 | - | 16 - 17 | 1612 | 65 |