Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AIFM3
Entrez Gene ID
150209
Gene Name
apoptosis inducing factor, mitochondria associated 3
Gene Aliases
AIFL
Chromosome Location
Chr.22: 20965130 - 20981360 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018060.2 NP_001018070.1 5 - 6 680 60
NM_001146288.1 NP_001139760.1 5 - 6 765 60
NM_144704.2 NP_653305.1 5 - 6 680 60
NR_027464.1 4 - 5 637 60
GenBank mRNA AK055035.1 - 5 - 6 664 60
AK295581.1 - 4 - 5 540 60
AK316042.1 - 4 - 5 533 60
BC032485.1 - 5 - 6 647 60
CR456342.1 - 4 - 5 499 60

View Full Product Details